This item is now discontinued. Please browse similar items
Vector Laboratories WestVision HRP Anti-Mouse IgG for Western Blot Detection
Representative image only.
Vector Laboratories Mfr # WB-2000-.8 – Item # WZ-93952-11
Improve your signal intensity without increasing background
  • Save time with ready-to-use format
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)0.8
  • Secondary AntibodyAnti-Mouse
  • ConjugatePeroxidase
  • Antibody HostHorse
  • Immunoglobulin ClassIgG
  • DescriptionHRP Anti-Mouse IgG for Western Blot Detection
  • Warranty1 year
1 Year Warranty
Key Features
  • Save time with ready-to-use format
  • 1 year warranty
More About this Item
WestVision™ Block and Diluent is a ready-to-use Tris buffer-based blocking solution intended for Western or dot blot applications. This solution is a proprietary formulation containing Tween 20 that improves signal intensity and resolution without increasing background. To achieve the best results, use WestVision™ Block and Diluent for all blocking and diluting steps including primary antibody and secondary antibody detection reagents. WestVision™ Block and Diluent is compatible with both alkaline phosphatase- and peroxidase- based detections systems, as well as with both chemiluminescence and chromogenic development.
Designed to detect primary antibodies of mouse or rabbit origin, the WestVision™ Peroxidase Polymer antibody conjugates produce a sharp signal and low background in western blot detection. These conjugates are based on a new method of polymerizing enzymes and attaching these polymers to antibodies. This allows for a higher density of enzymes to access a target with minimal interference. The result is higher sensitivity and a greater signal-to-noise ratio in western blot detection.