This item is now discontinued. Please browse similar items
Vector Laboratories PHOTOPROBE® Biotin (Long Arm) Reagent Kit
Representative image only.
Vector Laboratories Mfr # SP-1020-.5 – Item # WZ-93952-15
Preserves integrity of original sample
  • Labels DNA, RNA or PNA
  • Labels up to 250 μg of nucleic acid or up to 50 labeling reactions
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBiotin (Long Arm) Reagent Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Labels DNA, RNA or PNA
  • Labels up to 250 μg of nucleic acid or up to 50 labeling reactions
  • 1 year warranty
More About this Item
The PHOTOPROBE (Long Arm) Biotin reagent incorporates biotin at multiple sites along the entire length of the nucleic acid. It does not destroy the original nucleic acid or creates a copy. The labeling reagent has an extra-long linker arm to increase the distance between the sample and the tag.