This item is now discontinued. Please browse similar items
Variac Variable Autotransformer, Model . Single Phase, 120V, 50/60 Hz Input, Max mA 26.0, Open Frame
Representative image only.
Variac Mfr # 3090-5110 – Item # WZ-68865-45
Convenient, efficient, reliable load control
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input (VAC)120
  • Input (Hz)50/60
  • Output (amps)20
  • DescriptionVariable Autotransformer, Model . Single Phase, 120V, 50/60 Hz Input, Max mA 26.0, Open Frame
  • Warranty1 Year
1 Year WarrantyUL Listed.
Key Features
  • 1 year warranty
More About this Item
Applications –
Process control, instrumentation, aerospace and aviation,
defense, machine tool control, utilities, heat and light output,
robotics, power transmission
These autotransformers are efficient, trouble-free devices for controlling AC voltage and any other quantities that derive from AC voltage: heat output, light intensity, motor speed, and power supply outputs.
These continuously adjustable autotransformers are unlike most transformers. Units have a transformation ratio that can be smoothly and continuously changed so that the output of the unit can be controlled from zero to line voltage or even higher.