Vanguard Safety A16 Nitrile Exam Gloves, Large; 1000/CS
Representative image only.
Vanguard Safety Mfr # A16A13 – Item # WZ-78902-07
Maintain dexterity without sacrificing safety
  • Powder-free—made of soft 100% nitrile
  • Latex-free to eliminate adverse reactions
  • Textured surface for better gripping
  • Meets acceptable quality level (AQL)
  • Chemotherapy rated
  • FDA approved
  • Medical grade
  • Ambidextrous
  • Gloves are size large
PRODUCT OPTIONS
£85.65GBP / Case of 1000
£102.78GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5
  • SizeLarge
  • ColorBlue
  • GripTextured surface for improved grip
  • CuffAmbidextrous Beaded Cuff
  • SterileNo
  • Includes100 gloves/Box, 10 boxes/Case
  • PowderedNo
  • DescriptionNitrile Exam Gloves, Large; 1000/Cs
Key Features
  • Powder-free—made of soft 100% nitrile
  • Latex-free to eliminate adverse reactions
  • Textured surface for better gripping
  • Meets acceptable quality level (AQL)
  • Chemotherapy rated
  • FDA approved
  • Medical grade
  • Ambidextrous
  • Gloves are size large
More About this Item
These Vanguard Safety A16 nitrile exam gloves are made from powder-free and latex-free nitrile. They feature textured surface for improved grip and maximum ease of use with delicate instruments. All A16 nitrile gloves are ambidextrous and blue in color. Gloves are packed 100/box and 10 boxes per case.