This item is now discontinued. Please browse similar items
Vacuum Research 912093 Diaphragm Sensor, 30" Hg To Atm, 1/4" NPT (m)
Representative image only.
Vacuum Research Mfr # 912093 – Item # WZ-68808-66
Use with Hastings® DV-6 or VRC gauge tubes
  • Ranges from 10-5 Torr to 2 atmospheres
  • Analog output for PLC or computer interface
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Vacuum (\" Hg)30
  • Wetted Materials300 SS
  • Process Connection1/4\" NPT
  • Unit Of Measurement\" Hg
  • Media CompatibilityGases compatible with wetted materials
  • DescriptionDiaphragm Sensor, 30\" Hg To Atm, 1/4\" NPT (m)
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Ranges from 10-5 Torr to 2 atmospheres
  • Analog output for PLC or computer interface
  • 2 year warranty
More About this Item
These active vacuum switch/transmitters provide an inexpensive and straightforward way to control your vacuum system. Use the Pirani sensor switch directly to initiate crossover after roughing, or the diaphragm sensor switch to control backfill pressure on pump and purge operations. Feed the switch output into your PC or PLC and let the digital system make the control decisions based on the output signal.To create a complete vacuum system, choose your vacuum range and sensor type from the table below, and then choose your sensor connection style. Switch/transmitter plugs directly into sensors. Measure high, medium, and low vacuum with Pirani, thermocouple, and diaphragm sensors.