This item is now discontinued. Please browse similar items
Vacuubrand 2614455 Vacuu-Lan Mini-Network Vacuum Manifold, 3 valves
Representative image only.
Vacuubrand Mfr # 22614455 – Item # WZ-79210-01
Delivers vacuum down to 29.86" Hg
  • Completely modular, allowing you to extend capability one mini-network at a time
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithAny vacuum pump
  • DescriptionVacuu-Lan Mini-Network Vacuum Manifold, 3 valves
  • Warranty1 year
1 Year Warranty
Key Features
  • Completely modular, allowing you to extend capability one mini-network at a time
  • 1 year warranty
More About this Item
The VACUU-LAN mini-network provides three vacuum sources from one pump. To minimize interference among applications each individual port is check-valve-controlledreducing the risk of cross-contamination from other labs using a central vacuum system. Manual flow control at each port allows for variable speed adjustment. Used in conjuction with a vacuum pump . Combine with a vacuum pump on pages 0000-0000 for an instant filtration network.