This item is now discontinued. Please browse similar items
Uvex by Honeywell T5900WBLHS North Adaptec Safety Glasses, Dark Blue Frame, Smoke, HydroShield Antifog, Wide Lens; 10/BX
Representative image only.
Uvex by Honeywell Mfr # T5900WBLHS – Item # WZ-86498-04
Customized adjustability fits all faces
  • Smoke tint lens minimizes outdoor sunlight and glare HydroShield long-life antifog coating helps keep lenses clear Multi-adjustable frame design for a comfortable and snug fit
  • Dark Blue Frame Lens Size: Wide
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorDark Blue
  • Temple StyleAdjustable Tension
  • Lens ColorSmoke
  • Lens CoatingAntifog
  • Lens TypeWide
  • ComplianceANSI+ CSA Z94.3 (standard)
  • DescriptionSafety Glasses, Dark Blue Frame, Smoke, HydroShield Antifog, Wide Lens; 10/BX
Key Features
  • Smoke tint lens minimizes outdoor sunlight and glare HydroShield long-life antifog coating helps keep lenses clear Multi-adjustable frame design for a comfortable and snug fit
  • Dark Blue Frame Lens Size: Wide
More About this Item
With three different lens sizes to choose from, these scratch resistant polycarbonate vented glasses are sure to give you a more customized fit. They feature an adjustable lens angle and temple tension, along with a nose bridge, serving as added safeguards.
Compliance: Certified to meet ANSI+ CSA Z94.3 (standard).