This item is now discontinued. Please browse similar items
Uvex by Honeywell S3400HS Flex Seal Safety Goggles, Navy Body, Clear, HydroShield Antifog Lens; 1/EA
Representative image only.
Uvex by Honeywell Mfr # S3400HS – Item # WZ-86497-99
Conforms to almost any facial profile
  • Scratch-resistant, antifog and ideal for most indoor work environments Exceptionally snug and comfortable with soft body frame made of cured silicone Durable headband made of neoprene is resistant to oil, heat and weathering Pivoting clips allow for quick removal of headband
  • Navy Goggles
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorNavy
  • Lens ColorClear
  • Lens CoatingAntiscratch, Antifog
  • ComplianceANSI Z87.1-2015 and CSA Z94.3
  • DescriptionSafety Goggles, Navy Body, Clear, HydroShield Antifog Lens; 1/EA
Key Features
  • Scratch-resistant, antifog and ideal for most indoor work environments Exceptionally snug and comfortable with soft body frame made of cured silicone Durable headband made of neoprene is resistant to oil, heat and weathering Pivoting clips allow for quick removal of headband
  • Navy Goggles
More About this Item
It’s the silicone construction and design of these safety goggles that stand out from the rest. They provide all-day comfort, even when wearing them over prescription eyewear. Polycarbonate lens is clear and features an antiscratch and HydroShield antifog hard coating. Other lens tints for outdoor use, and a fabric headband is also available.
Compliance: ANSI Z87.1-2015 and CSA Z94.3 standards