Idex Flangeless Ferrule, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Mfr # P-300NX – Item # WZ-02007-56
No need to flange tubing
  • Excellent for connecting rigid or semi-rigid tubing
  • Natural ETFE
  • 10/PK
  • 1 month warranty
PRODUCT OPTIONS
£16.79GBP / Pkg of 10
£20.15GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • DescriptionFlangeless Ferrule, 1/8\", Tefzel, Pack Of 10
1 Month Warranty
Key Features
  • Excellent for connecting rigid or semi-rigid tubing
  • Natural ETFE
  • 10/PK
  • 1 month warranty
More About this Item
Use with male nuts for a leak-tight connection.
Flangeless Fittings are easy to assemble. Slide the ferrule over the tubing and fingertighten
the nut. Excellent chemical resistance. 1/8" fittings are rated to 500 psi; 1⁄16" fittings
are rated to 1000 psi. All fittings feature 1⁄4-28 threads.