This item is now discontinued. Please browse similar items
Uniweld Q4N85SM Refrigerant Manifold Sets, 4 Valve Aluminium manifold with 5 foot hose, 3-1/8"
Representative image only.
Uniweld Mfr # Q4N85SM – Item # WZ-07164-54
Manifolds for refrigerant
  • Brass Manifolds
  • Two easy-to-read gauges with color-coded refrigerant scales and boots
  • Includes 5 ft high-pressure hose set
  • 2-1/2'' gauges have limited pointer flutter
  • 3-1/8'' gauges have no pointer flutter on the manifolds
  • Extended vavles stems on the brass versions ensure adequate knuckle clearance
  • Reseatable PTFE valves stems packing for duability and leakproof reliability integrated in the brass versions
  • Aluminum Manifolds
  • Two easy-to-read gauges with color-coded refrigerant scales and boots
  • Includes 5 ft high-pressure hose set
  • No pointer flutter 3-1/8'' gauges
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRefrigerant Manifold Sets, 4 Valve Aluminium manifold with 5 foot hose, 3-1/8\"
  • Warranty1 year
1 Year Warranty
Key Features
  • Brass Manifolds
  • Two easy-to-read gauges with color-coded refrigerant scales and boots
  • Includes 5 ft high-pressure hose set
  • 2-1/2'' gauges have limited pointer flutter
  • 3-1/8'' gauges have no pointer flutter on the manifolds
  • Extended vavles stems on the brass versions ensure adequate knuckle clearance
  • Reseatable PTFE valves stems packing for duability and leakproof reliability integrated in the brass versions
  • Aluminum Manifolds
  • Two easy-to-read gauges with color-coded refrigerant scales and boots
  • Includes 5 ft high-pressure hose set
  • No pointer flutter 3-1/8'' gauges
  • 1 year warranty
More About this Item
Choose from brass or aluminum models.