This item is now discontinued. Please browse similar items
UltraTech 2317 Replacement Bladder for Ultra Spill Deck Systems
Representative image only.
UltraTech Mfr # 2317 – Item # WZ-86452-50
Hidden containment bladder automatically unfurls to capture large spills or drum leaks
  • Low profile, polyethylene containment decks meet EPA and other spill containment regulation
  • Compact footprint saves valuable floor space
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Bladder for Ultra Spill Deck Systems
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Low profile, polyethylene containment decks meet EPA and other spill containment regulation
  • Compact footprint saves valuable floor space
  • Limited Lifetime Warranty
More About this Item
By adding this 66-gallon bladder to any Spill Deck, we can provide a self-contained deck that meets the Federal Government's spill containment requirements. This unique, polyethylene bladder remains rolled up within deck—deploys only if spill occurs and allows for additional containment. This bladder is compatible with drums 86452-00, -02, and -04.
The Bladder will roll out 65.5" away from the deck and will be 66" wide when completely unfurled.