Aven Tools Tweezers, curved, Smooth Rounded Tips 2AB
Representative image only.
Aven Tools Mfr # 18.514 – Item # WZ-07290-06
£29.94GBP / Pkg of 12
£35.93GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 1/2
  • Overall Length (cm)11.43
  • MaterialPlastic
  • SterileNo
  • ESD SafeYes
  • DescriptionTweezers, curved, Smooth Rounded Tips 2AB
More About this Item
Ideal for handling semiconductors, silicone chips, and delicate wafers. Static dissipative for use with sensitive components where ESD damage is a concern. Temperature resistant up to 220°C; tips do not spread under pressure.