Troemner CLASS 7 1000g-1g Economical S.S. Class 7 Weight Set with Traceable Certificate

Representative image only.
Troemner Mfr # 301ST – Item # WZ-01029-26
This economical set is ideal for general purpose applications. Hinged polypropylene box with snap-tight closure.

PRODUCT OPTIONS

£235.00GBP / Each
£282.00GBP (incl VAT)
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Mass ClassClass 7
  • Mass CertificationNIST
  • Height (in)2.2812
  • Width (in)2.8281
  • Length (in)6.6562
  • Contents of Set1000g, 500g, 200g, (2) 100g, 50g, 20g, (2) 10g, 5g, (2) 2g, 1g; plastic weight block
  • Description1000g-1g Economical S.S. Class 7 Weight Set with Traceable Certificate

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat