Troemner 985690 Support Rod and Clamp Kit
Representative image only.
Troemner Mfr # 985690 – Item # WZ-84300-22
Effortlessly connect your probes or tubes to hotplate with support rod kit
  • Constructed of durable stainless steel
  • 2 year warranty
£211.11GBP / Each
£253.33GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSupport Rod and Clamp Kit
  • Warranty2 year
Two year warranty
Key Features
  • Constructed of durable stainless steel
  • 2 year warranty
More About this Item
Designed for holding temperature probes, thermometers or other glass tubes. Includes an 18" (45.7cm) stainless steel support rod, thermometer/temperature probe extension clamp, 3 prong medium swivel clamp, and hook connector.
Compatible with the Troemner Talboys basic mini hotplates, models 83400-02 and -04. Hot plate sold separately.