This item is now discontinued. We recommend the following as suggested replacements: WZ-11967-18

Troemner 7231-UCPW 50g-10 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate

Representative image only.
Troemner Mfr # 7231-UCPW – Item # WZ-01022-47
UltraClass Platinum -The weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0*. This class is used as a primary standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured. This class is appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001 mg.
  • UltraClass Platinum tolerance = ASTM Class 000
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
  • UltraClass Platinum weights come with a positive calibration tolerance gaurantee.

SPECS & DESCRIPTION

Specifications & Description

  • Mass ClassUltraClass Platinum
  • Mass Size (g)50
  • Mass CertificationNIST/NVLAP
  • Height (in)3.5
  • Width (in)7
  • Length (in)8
  • Contents of Set50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case
  • Description50g-10 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate

Key Features

  • UltraClass Platinum tolerance = ASTM Class 000
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
  • UltraClass Platinum weights come with a positive calibration tolerance gaurantee.

More About this Item

These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes. We guarantee for the life of all Troemner Alloy 8 Stainless Steel Precision Weights that they will maintain extremely low magnetic susceptibility.

Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.

Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat