This item is now discontinued. We recommend the following as suggested replacements: WZ-01032-32

Troemner 7221-UCGW 200g-1 mg, (2) Ultraclass Gold Mass Set, NVLAP Accredited Certificate

Representative image only.
Troemner Mfr # 7221-UCGW – Item # WZ-01022-66
UltraClass Gold - The weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1. This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured. This class is appropriate for calibrating high-precision analytical balances with a readability as low as 0.001 mg.
  • UltraClass Glold tolerance = ASTM Class 00
  • Weights include a Statement of Accuracy
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel

SPECS & DESCRIPTION

Specifications & Description

  • Mass ClassUltraClass Gold
  • Mass Size (g)200
  • Mass CertificationNIST/NVLAP
  • Height (in)3.5
  • Width (in)7
  • Length (in)8
  • Contents of Set(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case
  • Description200g-1 mg, (2) Ultraclass Gold Mass Set, NVLAP Accredited Certificate

Key Features

  • UltraClass Glold tolerance = ASTM Class 00
  • Weights include a Statement of Accuracy
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel

More About this Item

These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes.

Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.

Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat