ASTM Class 1 - Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as0. 1 mg to 0.01 mg .
Surfaces are polished to a perfect, mirror like finish
Masses are less vulnerable to wear and scratches
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
Description2g Analytical Class 1 Weight with NVLAP Accredited Certificate
Warranty3 month
Key Features
Surfaces are polished to a perfect, mirror like finish
Masses are less vulnerable to wear and scratches
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
3 month warranty
More About this Item
Appropriate for calibrating high-precision analytical balances with readability as low as 0.1 mg to 0.01 mg.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to,Manufacturing QC, Secondary Calibration Labs, State Laboratories, Pharmaceutical QC, R&D, Analytical Testing, Chemical Manufacturing, Utilities.