This item is now discontinued. Please browse similar items
Troemner 1274T 25Lb Class-F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
Representative image only.
Troemner Mfr # 1274T – Item # WZ-11966-44
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass F
  • Mass Size (lbs)25
  • Mass CertificationNIST/NVLAP
  • Description25Lb Class F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Stainless Steel Grip Handle Weights are legal for trade, which means their design meets specifications as outlined in NIST Handbook 105-1 and are approved to be used on legal for trade devices within the United States. Weights 5 kg/10 lb and below must be made of stainless steel of quality no lower than 300 series. Adjusting cavities are typically on the top of the weight and must be tamper resistant.
Stainless Steel Class F Test Weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable
for use in verifying scales that have 0.1% accuracy or lower. Class F Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Suggested markets for Stainless Steel Test Weights include, but are not limited to, Hospitals and Food Services.