This item is now discontinued. Please browse similar items

TriForest Boston Round Media Bottles, 500 mL, PP; 72/Cs

Representative image only.
TriForest Mfr # RPP0500 – Item # WZ-06542-29
Durable leakproof design for safe sample storage
  • Printed graduations for accurate measurement
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom that is ISO 9001 compliant
  • Autoclavable bottle and PP cap withstand repeated cycles
Often bought with…
Cole-Parmer® Essentials Glass Erlenmeyer Flasks, Graduated, Heavy Rim
£13.20 - £49.47GBP / Pkg of 8
£15.84 - £59.36GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialPolypropylene(PP)
  • Min Graduation (mL)500
  • Max Graduation (mL)50
  • Graduation Interval (mL)50
  • Cap MaterialPolypropylene
  • Cap / Stopper Size38-430
  • Cap ColorBlue
  • Bottle ColorTransparent
  • Withstands Low Temperature (° F)-40
  • Withstands High Temperature (° F)250
  • AutoclavableYes
  • Outside Diameter (OD) (in)3 1/5
  • Outside Diameter (OD) (cm)8.1
  • Overall Height (in)6 51/64
  • Overall Height (cm)17.2
  • DescriptionBoston Round Media Bottles, 500 mL, PP; 72/CS

Key Features

  • Printed graduations for accurate measurement
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom that is ISO 9001 compliant
  • Autoclavable bottle and PP cap withstand repeated cycles

More About this Item

Made of either polycarbonate (PC) or polypropylene (PP), these bottles are nonleaching and shatterproof, being a safer alternative to glass in storing media, buffers, reagents, and other aqueous solutions. Optically superior transparent bottles and blue caps make visual identification of samples quick and easy.
Bottles are provided nonsterile in a shrink-wrapped tray. The 500- and 1000-mL capacities have 12 bottles per tray and the 300-mL capacity has 24 bottles per tray.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat