This item is now discontinued. Please browse similar items
Trajan SGE eVoL Electronic Syringe Starter Kit; NMR Edition
Representative image only.
Trajan Mfr # 2910100 – Item # WZ-17940-24
World's first digital analytic syringes combine precision and speed
  • Program and store up to 98 steps of workflow
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Suitable for direct injection onto a chromatography column with consistent flow rate
  • Be safer when working with volatile samples — aliquots are dispensed into sealed vials
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeElectronic
  • Syringe Capacity (µL)500
  • Needle Tip StyleBevel
  • Needle Length (in)1.969
  • DescriptionEVOL Electronic Syringe Starter Kit; NMR Edition
Key Features
  • Program and store up to 98 steps of workflow
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Suitable for direct injection onto a chromatography column with consistent flow rate
  • Be safer when working with volatile samples — aliquots are dispensed into sealed vials
More About this Item
eVol XR syringes combine a digitally controlled electronic drive and XCHANGE® enabled analytics for improved accuracy and reproducibility. The eVol XR system allows quick and simple syringe changes, so syringes can be dedicated to types of samples or methods, helping prevent reagent cross-contamination. And calibration factors can be easily programmed and stored for each syringe, enabling compliance with global lab standards. Syringe programming can be password protected for security.
eVol XR Starter Kit for NMR includes 5 µL syringe with 11.5 cm needle, 50 µL syringe with 18 cm needle, and 500 µL syringe with 11.5 cm and 18 cm needles.
Available for sale in the Middle East, Europe, and United States only.
Available for sale in the Middle East, Europe, and United States only.