This item is now discontinued. Please browse similar items
Trajan SGE eVol® XR Digital Analytical Syringe, 50 µL, Removable Needle, 25G, Bevel Tip; 3/Pk
Representative image only.
Trajan Mfr # 2910322 – Item # WZ-17940-14
Digitally controlled electronic drive and an analytical syringe—all in one!
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Electronic syringe features a 50 µL capacity and 25G Bevel tip
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeDigital
  • Syringe Capacity (µL)50
  • Needle Tip StyleBevel
  • Needle IncludedYes
  • Needle Gauge25
  • Needle Length (in)2
  • MaterialStainless steel needle
  • SterileNo
  • DescriptionDigital Analytical Syringe, 50 µL, Removable Needle, 25G, Bevel Tip; 3/Pk
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Electronic syringe features a 50 µL capacity and 25G Bevel tip
More About this Item
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Available for sale in the Middle East, Europe, and United States only.