This item is now discontinued. We recommend the following as suggested replacements: WZ-94460-74
Traceable Sentry Triple-Display Thermometer with Calibration, Celsius; 1 Bottle Probe
Representative image only.
Traceable Item # WZ-94460-87
Simultaneously display high, low, and current temperature
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Monitors temperature continuously
  • Takes measurements in Celsius
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-62
  • Max Temperature (° F)161
  • Min Temperature (° C)-50
  • Max Temperature (° C)74
  • Resolution
  • Mounting TypeDesktop or wall-mount
  • Display Type1.25\" Display, 3.5-digit LCD
  • Height (in)2 1/2
  • Cable Length (in)120
  • Battery1.5v Battery (1)
  • DescriptionTraceable® Sentry Triple-Display Thermometer with Calibration; Celsius, Bottle Probe
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Monitors temperature continuously
  • Takes measurements in Celsius
  • 1 year warranty
More About this Item
Thermometers include a weatherproof remote sensor with a 10-ft. (3.05m) long 1/16"-diameter micro cable. The micro cable is ideal for applications that require a seal, such as a refrigerator, where a larger cable would interfere with the sealing of the door. Unplugging the external probe will activate an internal sensor for monitoring ambient temperature. Thermometer features a high-impact ABS plastic case, large LCD that updates every 10 seconds, and a simple one-button reset to clear the high/low memory. Supplied with hook-and-loop strip for wall mounting or has a flip-out stand for tabletop use.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Models 94460-86 and -87 are supplied with special a bottle probe specifically designed for refrigerator/freezer applications. The temperature sensor is enclosed in a bottle which protects it from rapid temperature changes when refrigerator door is opened.