This item is now discontinued. Please browse similar items
Traceable Remote Stainless Steel Sensor for raceable® Wireless Thermometer with Calibration
Representative image only.
Traceable Item # WZ-98767-23
Use with Traceable® Digital Wireless meters to accurately measure temperature and/or humidity
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithDigi-Sense Traceable Replacement Remote Sensor Module
  • Length (in)8.5
  • Diameter (in)0.125
  • Length (cm)21.6
  • Diameter (cm)0.3
  • DescriptionTraceable® Remote Stainless Steel Sensor for 94460-78 and 94460-79 with Calibration
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.