This item is now discontinued. Please browse similar items
Total Source APP0278-YELLOW Isolation barrier gowns, polyethylene-coated polypropylene, case of 50
Representative image only.
Total Source Mfr # APP0278-YELLOW – Item # WZ-86566-10
Disposable, low-cost gowns provide full frontal protection from particulates or light splashes
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)45.5
  • Length (cm)115.57
  • MaterialPolypropylene(PP)
  • SizeUniversal
  • ColorYellow
  • DescriptionIsolation barrier gowns, polyethylene-coated polypropylene, case of 50
More About this Item
Lightweight breathable gowns feature ties in back of neck and at waist for a secure, comfortable fit. Elastic cuffs at wrists keep the sleeves firmly in place and gather softly under gloves. Select 100% spunbound polypropylene blue gowns for durable, particulate protection. Select polyethylene-coated polypropylene yellow gowns for added protection from water based liquids and certain chemicals. Sold in case of 50.