Total Source APP0242-W Polyethylene-Coated Polypropylene Lab Coats, Small, case of 30
Representative image only.
Total Source Mfr # APP0242-W-S – Item # WZ-86227-10
Excellent lightweight splash protection
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)41.5
  • Length (cm)105.41
  • SizeSmall
  • MaterialPolyethylene(PE)
  • ColorWhite
  • DescriptionPolyethylene-Coated Polypropylene Lab Coats, Small
More About this Item
These polyethylene (PE) coated polypropylene (PP) lab coats are a low-cost disposable alternative to Tyvek® coats. The durable, lightweight, and breathable fabric is ideal for general-purpose laboratory applications. The PE-coated spun bond PP-provides excellent splash protection from water-based liquids.