This item is now discontinued. Please browse similar items
Torbal Roller Grip for Test Stand
Representative image only.
Torbal Mfr # 901106 – Item # WZ-25304-85
Various grips to suit your application needs
  • 3 years warranty parts and labor
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRoller Grip for Test Stand
  • Warranty3 years parts & labor
3 Year Warranty
Key Features
  • 3 years warranty parts and labor
More About this Item
Compatible with the Torbal STV Test Stand.
The roller grip attachment is used when measuring tape strength as well as tape adhesives. The grip is ideal for testing any sample that needs to be draped to obtain an accurate force measurement.