This item is now discontinued. Please browse similar items

Thermo Scientific STAR A325 Star A325 pH/Cond Portable Meter Only

Representative image only.
Thermo Scientific Mfr # STARA3250 – Item # WZ-58825-68
Advanced features and dependability makes it the top choice for measuring samples
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
  • Meets all US Pharmacopoeia (USP) requirements
  • Selectable cell constant for 2- or 4- cell conductivity probes
  • Selectable reference temperatures of 5,10, 15 20 or 25°C
  • 3 year warranty
Include Buffers Today!
Oakton® pH Buffer Solution Packs
£34.25 - £370.14GBP / Pkg of 3
£41.10 - £444.17GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeGraphic LCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Conductivity0 μS
  • Max Conductivity3000 mS
  • Conductivity Accuracy±0.5% FS + 1 LSD
  • Min TDS0 to 200 ppt
  • Min Salinity0 to 80 ppt
  • Resistivity Resolution2 Ω
  • Resistivity Accuracy±0.5% ±1 digit
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Battery4 x AA batteries (included)
  • Battery life (hr)800
  • Width (cm)10.795
  • Height (cm)23.1775
  • Width (in)4 1/4
  • Length (in)2 3/8
  • Height (in)9 1/8
  • DescriptionStar A325 pH/Cond Portable Meter Only
  • Warranty3 year
3 Year WarrantyIP 67

Key Features

  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
  • Meets all US Pharmacopoeia (USP) requirements
  • Selectable cell constant for 2- or 4- cell conductivity probes
  • Selectable reference temperatures of 5,10, 15 20 or 25°C
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A322 conductivity and A325 pH/conductivity meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Available as meter only and as complete meter kit configurations

Meter only includes four AA batteries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat