Thermo Scientific STAR A211 Star A211 pH Benchtop Meter Kit

Representative image only.
Thermo Scientific Mfr # STARA2115 – Item # WZ-58825-24
Durability and dependability make it the ideal choice for every lab, every sample, every day
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
  • 3 year warranty

£1,403.19GBP / Each
£1,683.83GBP (incl VAT)

£187.00 - £208.21GBP / Each
£224.40 - £249.85GBP (incl VAT)

for an additional
£187.00GBP / Each
£224.40GBP (incl VAT)
for an additional
£208.21GBP / Each
£249.85GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Oakton Electrode Storage Solution; 500 mL
£21.65GBP / Pint of 1
£25.98GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature Resolution0.1
  • Temperature Accuracy±0.1°C
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232 and USB
  • Data Logging (points)2000 data points
  • WaterproofYes
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • Battery life (hr)800
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionStar A211 pH Benchtop Meter Kit
  • Warranty3 year
3 Year WarrantyIP54 Rated

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A211 pH, mV, RmV and temperature meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

Up to five point automatic calibration with USA/NIST and DIN buffers, auto-buffer recognition buffers, and recalibration feature to fix calibration errors. GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Meter kit includes: ROSS Triode pH/ATC electrode (05718-28), 475 mL each of 4, 7, 10 pH buffer and electrode storage solution, 60 mL of cleaning solution, swing-arm electrode holder and universal power adapter.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat