This item is now discontinued. Please browse similar items

Thermo Scientific Owl P2-20G Replacement blank glass plate for model 28554-15

Representative image only.
Thermo Scientific Owl Mfr # P2-20G – Item # WZ-28554-56
Optimize performance of your electrophoresis system

SPECS & DESCRIPTION

Specifications & Description

  • Glass Width (in)7.87
  • Glass Width (cm)20
  • Glass Height (cm)20
  • Thickness (in)0.1875
  • Thickness (cm)0.48
  • DescriptionReplacement blank glass plate for model 28554-15

More About this Item

Choose from a wide range of accessories for your Thermo Scientific Owl Large-Vertical Electrophoresis System 28554-15

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat