Thermo Scientific Nunc Lab-Tek™ Petri Dishes, Sterile, Polystyrene, Square, Vented, 100 x 15 mm; 500/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 4021 – Item # WZ-14007-10
Get better cultures
  • Ideal for fungi, bacteria, and other microorganisms
  • Sterilized by gamma irradiation
PRODUCT OPTIONS
£1,184.51GBP / Case of 500
£1,421.41GBP (incl VAT)
Parafilm PM992 M Wrap, 2
£37.84GBP / Roll of 1
£45.41GBP (incl VAT)
Consider Always in Stock Alternative
SPECS & DESCRIPTION
Specifications & Description
  • Well Diameter (mm)100
  • Well ShapeSquare
  • Culture Area (cm²)79
  • Surface TreatmentNone
  • Lid (Included)Yes, Vented
  • SterileYes
  • Height (mm)15
  • DescriptionPetri Dishes, Sterile, Polystyrene, Square, Vented, 100 x 15 mm; 500/Cs
Key Features
  • Ideal for fungi, bacteria, and other microorganisms
  • Sterilized by gamma irradiation
More About this Item
These sterile and disposable polystyrene (PS) dishes are ideal for culturing fungi, bacteria, and other microorganisms. Dishes work well in robotic dispensers due to flatness and uniform height. Sterilized by gamma irradiation.