This item is now discontinued. Please browse similar items
Thermo Scientific Nunc 2650-0100 Cylindrical Closed-Dome Tank, PP, 100 Gal.
Representative image only.
Thermo Scientific Nunc Mfr # 2650-0100 – Item # WZ-50301-56
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Wall thickness (in)0.312
  • Volume (Gal)100
  • Volume (Liters)379
  • Height (in)51 1/4
  • Height (cm)130.175
  • Outside Diameter (OD) (in)28 1/2
  • DescriptionCylindrical Closed-Dome Tank, PP, 100 Gal.
More About this Item
The Polypropylene Closed-Dome Tank has a 30-gallon capacity. Mount mixers off-center—a baffle is not required for optimum mixing. Tanks are constructed of polypropylene. You must order sanitary mixer mount 50301-70 with the tank. Cover is included.
Thermo Scientific Nalgene—Reg TM Nalge Thermo Scientific Nunc International