This item is now discontinued. Please browse similar items
Thermo Scientific Nunc 2650-0055 Cylindrical Closed-Dome Tank, PP, 55 Gal.
Representative image only.
Thermo Scientific Nunc Mfr # 2650-0055 – Item # WZ-50301-54
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Wall thickness (in)0.25
  • Volume (Gal)55
  • Volume (Liters)208
  • Height (in)39 1/2
  • Height (cm)100.33
  • Outside Diameter (OD) (in)22
  • DescriptionCylindrical Closed-Dome Tank, PP, 55 Gal.
More About this Item
The Polypropylene Closed-Dome Tank has a 30-gallon capacity. Mount mixers off-center—a baffle is not required for optimum mixing. Tanks are constructed of polypropylene. You must order sanitary mixer mount 50301-70 with the tank. Cover is included.
Thermo Scientific Nalgene—Reg TM Nalge Thermo Scientific Nunc International