Thermo Scientific Nunc Cell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 4-Well, 1 mL; 120/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 179820 – Item # WZ-01928-60
Non-treated surface offers low cell attachment
  • Certified nonpyrogenic and sterile
  • Excellent optical properties
  • Raised well rims to lower the risk of cross-contamination
  • Lids feature notched corners for good fit and easy orientation
  • Overall dimensions: 66 x 66 cm
PRODUCT OPTIONS
£422.39GBP / Case of 120
£506.87GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells4
  • Max Working Well Volume (mL)1
  • Well ShapeFlat
  • Culture Area (cm²)500
  • Surface TreatmentNone
  • Lid (Included)Yes
  • SterileYes
  • DescriptionCell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 4-Well, 1 mL; 120/Cs
Nonpyrogenic
Key Features
  • Certified nonpyrogenic and sterile
  • Excellent optical properties
  • Raised well rims to lower the risk of cross-contamination
  • Lids feature notched corners for good fit and easy orientation
  • Overall dimensions: 66 x 66 cm
More About this Item
These sterile polystyrene tissue culture dishes are used for unique cell growth applications that do not require the Nunclon Delta surface. Non-treated surface offers low cell attachment. Clear, uniform design provides excellent optical quality, making it a suitable choice for microscopy.