Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 75 cm2; 100/ Cs
Representative image only.
Thermo Scientific Nunc Mfr # 156472 – Item # WZ-01930-49
Nunclon™ Delta surface treatment provides optimal cell attachment and growth
  • EasYFlask™ ergonomic design features a 1/3 turn to open and close the cap
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
  • Solid caps feature a “Y” mark that allows visual verification of vent position
PRODUCT OPTIONS
£461.22GBP / Case of 100
£553.46GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeVented
  • Cap MaterialHDPE
  • Neck TypeAngled
  • Working Volume (mL)25
  • Culture Area (cm²)75
  • DescriptionCell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 75 cm2; 100/ Cs
Nonpyrogenic
Key Features
  • EasYFlask™ ergonomic design features a 1/3 turn to open and close the cap
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
  • Solid caps feature a “Y” mark that allows visual verification of vent position
More About this Item
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. Low profile design maximizes incubator space. Flasks feature volume graduations printed and molded on both sides and slightly angled side walls allow for complete viewing under a microscope.
Vented caps feature a set vent position that produces a finger-felt positive stop when fully open for gas exchange and the Y on the cap provides a visual indication of the vent/close position when stacked in incubators. Filtered caps allow constant airflow and minimize chances of contamination.