Thermo Scientific Nalgene Pipette Cleaning Basket for 16" L Pipets
Representative image only.
Thermo Scientific Nalgene Mfr # DS5241-0040 – Item # WZ-06150-00
Protect glass pipets from breakage while cleaning
  • Constructed of durable HDPE
  • For use with Thermo Scientific™ Nalgene™ pipette jars and washers
  • Use with pipette jar 06172-00 and pipette washer 06185-00
  • 1 year warranty
PRODUCT OPTIONS
£275.66GBP / Each
£330.79GBP (incl VAT)
Pipette Washer...
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)22 3/4
  • Wetted MatterialHDPE
  • Compatible withPipette jar 06172-00 and pipette washer 06185-00
  • DescriptionPipette Cleaning Basket for 16" L Pipets
1 Year Warranty
Key Features
  • Constructed of durable HDPE
  • For use with Thermo Scientific™ Nalgene™ pipette jars and washers
  • Use with pipette jar 06172-00 and pipette washer 06185-00
  • 1 year warranty
More About this Item
Protect glass pipets from breakage by using resilient, chemical-resistant Thermo Scientific™ Nalgene™ HDPE Pipet Baskets. A perforated screen in the bottom allows cleaning solutions to fill the chamber from the bottom and is recessed 0.75″ (1.9cm) for complete drainage. Use these high-denisty polyethylene baskets inside Nalgene pipet jars to soak pipets in cleaning or disinfection solutions, then drain and transfer into the Nalgene pipet washer for automatic pipet washing and rinsing.