Thermo Scientific Nalgene Cryogenic Vial Colored Cap Insert, Polycarbonate, Blue; 100/Pk
Representative image only.
Thermo Scientific Nalgene Mfr # 5045-0003 – Item # WZ-06755-94
Color-code your cryogenic vials with these cap inserts for quick identification
  • Constructed of durable polycarbonate
PRODUCT OPTIONS
£38.71GBP / Pkg of 100
£46.45GBP (incl VAT)
Argos Technologies PolarSafe® Cryovial Cap Insert, Blue; 100/Pk
£7.26GBP / Pkg of 100
£8.71GBP (incl VAT)
Often Bought With
Thermo Scientific™ Nalgene™ Cryogenic Vials, External Thread, Sterile
£23.52 - £56.00GBP / Pkg of 10
£28.22 - £67.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeCap Insert
  • Cap MaterialPolycarbonate
  • Cap colorBlue
  • DescriptionCryogenic Vial Colored Cap Insert, Polycarbonate, Blue; 100/Pk
Key Features
  • Constructed of durable polycarbonate
More About this Item
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.