Thermo Scientific Nalgene Filling/Venting Cap for 1/2" Tubing, 83B
Representative image only.
Thermo Scientific Nalgene Mfr # 2162-0830 – Item # WZ-06258-00
Transfer aseptic liquid safely
  • Autoclavable
  • 1 year warranty
PRODUCT OPTIONS
£92.42GBP / Each
£110.90GBP (incl VAT)
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID12.7 mm
  • Descriptionfilling/venting cap for 1/2\" tubing, 83B
  • Warranty1 year
1 Year Warranty
Key Features
  • Autoclavable
  • 1 year warranty
More About this Item
Filling/venting caps have thermoplastic elastomer (TPE) gaskets to allow aseptic liquid transfer.
Include 6" (15 cm) and 30" (76 cm) lengths of platinum-cured silicone tubing.