This item is now discontinued. Please browse similar items
Thermo Scientific Hypersil BDS C8 Column - Length 150mm, 4.6mm ID, 3µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 28203-154630 – Item # WZ-01885-94
Long life and trusted reproducibility for improved analyses
  • pH stable from 2 to 9
  • Base deactivating and endcapping reduce peak tailing for more precise results
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctylsilane (C8)
  • Column Particle Size (µm)3
  • DescriptionBDS C8 Column - Length 150mm, 4.6mm ID, 3µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • pH stable from 2 to 9
  • Base deactivating and endcapping reduce peak tailing for more precise results
  • 1 month warranty
More About this Item
Thermo Scientific Hypersil BDS (Base Deactivated Silica) columns have been trusted worldwide for over 20 years. Each column is manufactured with the same exactness and come with a certificate of analysis, ensuring you that each column will perform as promised. These high standards of manufacturing provide you with consistency and reliability year after year, meaning that you will not have to worry about changing procedures when you need to replace your column.
Hypersil BDS C8 Columns provide reliability for routine analysis, including acids, bases, and neutral compounds. These columns offer similar selectivity as the C18, with slightly less retention.