This item is now discontinued. Please browse similar items
Thermo Scientific Hypersil 25802-152130 GOLD CN Column - Length 150mm, 2.1mm ID, 1.9µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 25802-152130 – Item # WZ-01883-90
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseCyano
  • Column Particle Size (µm)1.9
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionGOLD CN Column - Length 150mm, 2.1mm ID, 1.9µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Thermo Scientific Hypersil GOLD CN Columns provide excellent peak shape and selectivity in reversed and normal phase separations, with lower hydrophobicity when compared to alkyl chain phases. These columns extend the range of available selectivities, and can be used in the analysis of compounds including steroids. Stable in a pH range of 2 to 8.