This item is now discontinued. Please browse similar items
Thermo Scientific Hypersil 25202-152130 GOLD C8 Column - Length 150mm, 2.1mm ID, 1.9µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 25202-152130 – Item # WZ-01879-23
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctylsilane (C8)
  • Column Particle Size (µm)1.9
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionGOLD C8 Column - Length 150mm, 2.1mm ID, 1.9µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Thermo Scientific Hypersil GOLD C8 Columns provide a similar selectivity to the Hypersil GOLD columns but with less retention and a predictable elution order. By reducing the hydrophobic interactions within the column, the C8 columns are able to elute compounds more quickly. Stable in a pH range of 2 to 9.