This item is now discontinued. Please browse similar items
Thermo Scientific Hypersil 25003-152130 GOLD Column - Length 150mm, 2.1mm ID, 3µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 25003-152130 – Item # WZ-01877-75
Achieve exceptional peak shake and resolution of your HPLC and LC/MS application
  • Significant reduction in peak tailing
  • Excellent resolution, efficiency, and sensitivity
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSilica
  • Column Particle Size (µm)3
  • DescriptionGOLD Column - Length 150mm, 2.1mm ID, 3µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • Significant reduction in peak tailing
  • Excellent resolution, efficiency, and sensitivity
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil GOLD™ C18 Selectivity HPLC Columns are silica-based and contain an even surface, reducing unwanted secondary and tertiary interactions. This reduction of interactions reduces peak tailing while maintaining C18 selectivity. The excellent peak symmetry leads to improved sensitivity, allowing for easier peak integration calculations. Stable in a pH range of 1 to 11.