Thermo Scientific GENESYS™ Long Path 20 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
Representative image only.
Thermo Scientific Mfr # 840-277700 – Item # WZ-02654-62
Enhance the versitility of your spectrophotometer
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
PRODUCT OPTIONS
£180.53GBP / Each
£216.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithGENESYS 30, 40, and 50 spectrophotometers
  • DescriptionLong Path 20 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
  • Warranty2 year
Two year warranty
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Long path rectangular cell holder is suitable for 10 to 100 mm pathlength cuvettes.
Available for sale and use in the United States only.