This item is now discontinued. Please browse similar items

Thermo Scientific BioBasic 73205-054030 SCX Column - Length 50mm, 4.0mm ID, 5µm particle size

Representative image only.
Thermo Scientific BioBasic Mfr # 73205-054030 – Item # WZ-01901-70
Stable polymeric coating suitable for separation and retention of basic and other cationic species
  • 1 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • PhaseSCX
  • Column Length (mm)50
  • Column Inside Diameter (ID) (mm)4
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4
  • DescriptionSCX Column - Length 50mm, 4.0mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty

Key Features

  • 1 month warranty

More About this Item

Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic SCX Columns utilize a polymeric sulfonic acid ion exchange ligand that is bound to a highly pure, base-deactivated silica that creates a stable polymeric coating, even in demanding pH conditions (pH range of 2 to 8). Proven in the separation of cationic proteins and peptides, as well as neurotransmitters, vitamins, and quaternary amine herbicides. These columns are extremely reliable, providing reproducible results run after run.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat