This item is now discontinued. Please browse similar items
Thermo Scientific BioBasic 72205-101030 8 Column - Length 100mm, 1.0mm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 72205-101030 – Item # WZ-01898-05
Provide excellent separation of a wide range of peptides
  • Excellent starting column for peptide and protein separations
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctylsilane (C8)
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)1
  • Description8 Column - Length 100mm, 1.0mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • Excellent starting column for peptide and protein separations
  • 1 month warranty
More About this Item
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic 8 Columns contain 300Å pore size silica, which is ideally suited for medium-hydrophobicity peptides nominally less than 20,000 Da. Providing less hydrophobic retention than BioBasic 18 columns, BioBasic 8 columns are great starting columns for protein separations and may provide changes in elution patterns. Columns provide a highly stable, reproducible surface with sharp peak shap