This item is now discontinued. Please browse similar items
Thermo Scientific BioBasic 72105-051564 18 Nanobore Column with IntegraFrit - 150µm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 72105-051564 – Item # WZ-01897-22
Provide high efficiency and sensitivity – ideal for LC/MS analysis
  • KAPPA Capillary Columns are especially suitable for small sample, proteomic separations
  • Nanobore Columns provide higher sensitivity for nanospray applications
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseOctadecyl silane (C18)
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)0.15
  • Description18 Nanobore Column with IntegraFrit - 150µm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • KAPPA Capillary Columns are especially suitable for small sample, proteomic separations
  • Nanobore Columns provide higher sensitivity for nanospray applications
  • 1 month warranty
More About this Item
BioBasic Nanobore Columns are available in internal diameters of 75µm and 150µm. The nanobore columns are ideal for nanospray, LC/MS applications, particularly proteomics. Low flow rates (nL/min) offer a higher sensitivity with greater signal-to-noise ratio than traditional electrospray. The nanobore columns with IntegraFrit incorporate a high-porosity frit, with the chromatography bed packed behind this frit. Nanobore columns with PicoFrit spray directly from the column, eliminating performance losses and increasing MS performance. Manufactured from inert tubing and unions compatible wuth 10 - 32 male nuts and 1/16" OD capillary tubing.