This item is now discontinued. Please browse similar items
Thermo Scientific Barnstead D8951 Ion Exchange Cartridge; 2/3-Size, Mixed-Bed
Representative image only.
Thermo Scientific Barnstead Mfr # D8951 – Item # WZ-99260-05
Customize your system with application-specific water purification cartridges
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MediaCombination resin
  • Capacity (grains)470 as Sodium Chloride
  • Connection3/8\" hose barb at each end
  • FunctionIon exchange with activated carbon
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge; 2/3-Size, Mixed-Bed
  • Warranty2 year
Two year warranty
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Color-changing resin indicates when cartridge is exhausted. Cannot be used with alcohol-containing fluids.