This item is now discontinued. Please browse similar items
Thermo Scientific Barnstead D50254 Type I Cartridge, organic free, Nanopure Infinity
Representative image only.
Thermo Scientific Barnstead Mfr # D50254 – Item # WZ-99275-82
Keep systems operating precisely
  • For use with Nanopure Infinity systems
  • Include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water
  • Organic-free cartridge produces water that has less than 10 ppb organic carbin
  • Includes final filter
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible withThermo Scientific Barnstead Type I
  • DescriptionType I Cartridge, organic free, Nanopure Infinity
  • Warranty1 year
1 Year Warranty
Key Features
  • For use with Nanopure Infinity systems
  • Include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water
  • Organic-free cartridge produces water that has less than 10 ppb organic carbin
  • Includes final filter
  • 1 year warranty
More About this Item
Kit contains one 99275-61 (pretreatment), two 99275-62 (ultrapure), one 99275-63 (organic free), and two 99261-60 (final filter).