This item is now discontinued. Please browse similar items
Thermo Scientific Barnstead 50132196 LabTower TII 40 l/hr with 100 L tank
Representative image only.
Thermo Scientific Barnstead Mfr # 50132196 – Item # WZ-99290-54
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ASTM Water GradeType II
  • Capacity40 L/hr
  • Inlet PortR 3/4\"
  • With UV LampYes
  • Feed Water Requirements (Source)tap water, softened or hardness stabilized
  • Feed Water Requirements (Conductivity)<1500 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Type II Water (Resistivity)15 to 10 MΩ-cm
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type II Water (TOC)<30 ppb
  • Type II Water (Conductivity)0.067 to 0.1 µS-cm
  • DescriptionLabTower TII 40 l/hr with 100 L tank
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
The Barnstead LabTower TII system delivers up to 240L/day of Type 2 pure water (10-15 megohm) from a tap feed water supply. These units are available in 2 product water output sizes, 20 LPH or 40LPH.
LabTower TII system applications requiring large volumes of type 2 water such as reagent preparation, media preparation and glassware rinsing can be met.
LabTower TII system standard features include dispensing of type 2 water from the integrated 100L reservoir or from the system, quick connect cartridges, universal power supply, variable flow control knob, reservoir level controller, pressure regulator, RS232, and core consumables included with the system. Optional features include choice of systems with or without built in UV lamp, external printer, and IOQ documentation (order each separately).