This item is now discontinued. Please browse similar items
Thermo Scientific Barnstead 50132125 Pacific TII Water Purification Systems 20 l/hr
Representative image only.
Thermo Scientific Barnstead Mfr # 50132125 – Item # WZ-99290-18
Multiple capacities available to support your daily requirements
  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ASTM Water GradeType II
  • Max Production (L/hour)20
  • Max Flow Rate (L/hour)20
  • Inlet PortR 3/4\"
  • With UV LampNo
  • Feed Water Requirements (Source)tap water, softened or hardness stabilized
  • Feed Water Requirements (Conductivity)<1500 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Type II Water (Resistivity)15 to 10 MΩ-cm
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type II Water (TOC)<30 ppb
  • Type II Water (Conductivity)0.067 to 0.1 µS-cm
  • DescriptionPacific TII Water Purification Systems 20 l/hr
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty
More About this Item
Systems deliver up to 240 L/day of Type II pure water from a tap feed water supply. Ideal for routine lab work such as reagent preparation, media preparation, and glassware rinsing. High performance reverse osmosis membrane removes approximately 98% of inorganic ions and 99% of all dissolved organic substances, as well as micoorganisms and particles.
Space-saving design allows benchtop operation or wall mounting. Large, illuminated four-line display is easy to read and clearly shows operating mode status (production, standby, disinfection), reservoir volume (%), and temperature (°C). Display can be tilted for optimal viewing.
Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately. Optional UV lamp (254 nm) acts as a germicide, preventing biological contamination.
Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection. Reservoirs are available with an optional recirculation pump which protects high purity water from bacterial growth during standstill. The 30- and 60-L units can be used with the optional wall-mounting bracket for further space-saving capabilities.