This item is now discontinued. Please browse similar items
Thermo Scientific AQUAFAST Aquafast Nutrient Test Kit , Ammonia, 0.02 to 2.5 mg/L, 50/Pk
Representative image only.
Thermo Scientific Mfr # ACR012 – Item # WZ-99570-20
  • Each kit includes 50 vials with pre-measured reagents
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersAmmonia
  • Number of Tests Included50
  • Range0.02 to 2.5 mg/L
  • DescriptionAquafast Nutrient Test Kit , Ammonia, 0.02 to 2.5 mg/L, 50/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Each kit includes 50 vials with pre-measured reagents
  • 1 year warranty
More About this Item
Kits require a reactor for digestion (99581-35), and a colorimeter to read results.